Learn More
Biomatik Corporation Human Sulfatase 2 (SULF2) CLIA Kit
Supplier:  Biomatik Corporation EKU09899
Encompass_Preferred
Human Sulfatase 2 (SULF2) CLIA Kit for detecting SULF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.14-100ng/ml. Sensitivity: 0.09ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q8IWU5. Gene ID: 55959. Target Synonyms: HSULF-2; Extracellular sulfatase Sulf-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6247