Learn More
Biomatik Corporation Human Sulfatase Modifying Factor 1 (SUMF1) ELISA Kit
Supplier:  Biomatik Corporation EKU07495
Encompass_Preferred
Human Sulfatase Modifying Factor 1 (SUMF1) ELISA Kit for detecting SUMF1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8NBK3. Gene ID: 285362. Target Synonyms: FGE; C-alpha-formylglycine-generating enzyme 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5570