Learn More
Sino Biological Human SULT2B1 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786370
Encompass_Preferred
The Human SULT2B1 ELISA Pair Set is for the quantitative determination of Human SULT2B1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for SULT2B1 coated on a 96-well plate. Standards and samples are added to the wells, and any SULT2B1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-SULT2B1 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of SULT2B1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86370 Supplier Sino Biological Supplier No. SEK1141015
The Human SULT2B1 ELISA Pair Set is for the quantitative determination of Human SULT2B1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for SULT2B1 coated on a 96-well plate. Standards and samples are added to the wells, and any SULT2B1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-SULT2B1 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of SULT2B1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.