Learn More
Sino Biological Recombinant Anti-SUMO1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 13095R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SUMO1 / SUMO-1 (rh SUMO1 / SUMO-1; Catalog#13095-H07E; AAH66306.1; Ser2-Val101).
Catalog No. 50-255-4315