Learn More
Sino Biological Anti-SUMO1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13095RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SUMO1 / SUMO-1 (rh SUMO1 / SUMO-1; Catalog#13095-H07E; AAH66306.1; Ser 2-Val 101). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4314