Learn More
Sino Biological Human SUMO1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13095RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SUMO1 / SUMO-1 (rh SUMO1 / SUMO-1; Catalog#13095-H07E; AAH66306.1; Ser 2-Val 101). SUMO1 / SUMO-1 specific IgG was purified by Human SUMO1 / SUMO-1 affinity chromatography.
Catalog No. 50-237-3489