Learn More
Sino Biological Recombinant Human SUMO1 Protein (His Tag) 500
Supplier:  Sino Biological 13095H07E500
Encompass_Preferred
A DNA sequence encoding the human SUMO1 (AAH66306.1) (Ser 2-Val 101) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-224-6923