Learn More
Biomatik Corporation Human Suppressor Of Zeste 12 Homolog (SUZ12) ELISA Kit
Supplier:  Biomatik Corporation EKU07512
Encompass_Preferred
Human Suppressor Of Zeste 12 Homolog (SUZ12) ELISA Kit for detecting SUZ12 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15022. Gene ID: 23512. Target Synonyms: CHET9; JJAZ1; Chromatin precipitated E2F target 9 protein; Joined to JAZF1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8746