Learn More
Biomatik Corporation Human Survival Of Motor Neuron 2, Centromeric (SMN2) ELISA Kit
Supplier:  Biomatik Corporation EKU07533
Encompass_Preferred
Human Survival Of Motor Neuron 2, Centromeric (SMN2) ELISA Kit for detecting SMN2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16637. Gene ID: 6606. Target Synonyms: SMNC; SMNT; SMA2; BCD541; SMA3; SMA4; SMA1; SMA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4341