Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SUZ12 (aa 389-526) Control Fragment Recombinant Protein

Catalog No. RP102304
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP102304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP102304 Supplier Invitrogen™ Supplier No. RP102304
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84345. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SUZ12 (polycomb protein SUZ12) is a zinc finger gene and has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. SUZ12 is also a component of the PRC2/EED-EZH2 complex which methylates Lys-9 (H3K9me) and Lys-27 (H3K27me) of histone H3 leading to transcriptional repression of the affected target gene. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15022
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 23512
Name Human SUZ12 (aa 389-526) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2610028O16Rik; AI195385; AU016842; AW536442; chET 9 protein; CHET9; chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; Kiaa0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog; suppressor of zeste 12 homolog (Drosophila); suppressor of zeste 12 protein homolog; Suz12; SUZ12 polycomb repressive complex 2 subunit
Common Name SUZ12
Gene Symbol SUZ12
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NKPGSVKPTQTIAVKESLTTDLQTRKEKDTPNENRQKLRIFYQFLYNNNTRQQTEARDDLHCPWCTLNCRKLYSLLKHLKLCHSRFIFNYVYHPKGARIDVSINECYDGSYAGNPQDIHRQPGFAFSRNGPVKRTPIT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.