Learn More
Biomatik Corporation Human Synaptophysin (SYP) ELISA Kit
Supplier:  Biomatik Corporation EKU10232
Encompass_Preferred
Human Synaptophysin (SYP) ELISA Kit for detecting SYP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.5-10,000pg/ml. Sensitivity: 53.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P08247. Gene ID: 6855. Target Synonyms: Major synaptic vesicle protein p38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8360