Learn More
BIOHIPPO Human SYNC Microsample
SDP
Catalog No. 5000314509
Encompass
Human SYNC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SYNC in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 113 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SYNC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SYNC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SYNC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14509 Supplier BIOHIPPO Supplier No. BHE15211585
Human SYNC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SYNC in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 113 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SYNC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SYNC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SYNC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color