Learn More
Biomatik Corporation Human Syncoilin (SYNC) ELISA Kit
Supplier:  Biomatik Corporation EKU07546
Encompass_Preferred
Human Syncoilin (SYNC) ELISA Kit for detecting SYNC in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H7C4. Gene ID: 81493. Target Synonyms: Syncoilin-1; Intermediate Filament Protein; Syncoilin intermediate filament 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7246