Learn More
ACROBiosystems Human Syndecan-1 Protein, Fc Tag
Supplier: ACROBiosystems SY1H5253100UG
Recombinant Protein;100UG;Human Syndecan-1, Fc Tag (SY1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.3 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Syndecan-1, Fc Tag (Cat. No. SY1-H5253) at 1 μg/mL (100 μL/well) can bind Anti-Syndecan-1 antibody, Mouse IgG1 with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Anti-Syndecan-1 antibody, Mouse IgG1 at 1 μg/mL (100 μL/well) can bind Human Syndecan-1, Fc Tag (Cat. No. SY1-H5253) with a linear range of 1-2 ng/mL (Routinely tested).