Learn More
Abcam Human Syntaxin 1B ELISA Kit, 1 x 96 Tests
Catalog No. 502711638
Encompass_Preferred
Human Syntaxin 1B ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Syntaxin 1B with a sensitivity of 16.949 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-1638 Supplier Abcam Supplier No. AB3169011X96TESTS
Add to Cart
Add to Cart
Human Syntaxin 1B ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Syntaxin 1B with a sensitivity of 16.949 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement