Learn More
Biomatik Corporation Human Syntaxin 2 (STX2) ELISA Kit
Supplier:  Biomatik Corporation EKU07557
Encompass_Preferred
Human Syntaxin 2 (STX2) ELISA Kit for detecting STX2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.109ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32856. Gene ID: 2054. Target Synonyms: EPIM; EPM; STX2A; STX2B; STX2C; Epimorphin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5471