Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human T Cell Backbone Panel Kit, BD Horizon™

Catalog No. BDB568263 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568263 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB568263 Supplier BD Biosciences Supplier No. 568263
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

BV786 Mouse Anti-Human CD4: 100 Tests (1 ea) : R718 Mouse Anti-Human CD8: 100 Tests (1 ea) : PE-Cy7 Mouse Anti-Human CD45RA: 100 Tests (1 ea) : BV510 Mouse Anti-Human CD3: 100 Tests (1 ea) : BV711 Mouse Anti-Human CCR7 (CD197): 100 Tests (1 ea) : Brilliant Stain Buffer: 100 Tests (1 ea)

Antibody Panel Kit

The Human T Cell Backbone Panel contains five individual vials of fluorochrome-conjugated antibodies against T cell core markers [CD3, CD4, CD8, CD45RA, CD197 (CCR7)] conventionally used to identify different maturational states of CD4+ and CD8+ T cells (naïve, central memory, effector memory, effector memory RA). The kit also contains the BD HorizonTM Brilliant Stain Buffer for optimal performance. The Human T Cell Backbone Panel is strategically designed to be complemented with 4-5 drop-in fluorochromes, ie, fluorescent reagents specific for biomarkers of choice, depending on instrument configuration, with minimal panel design effort. Assignment of fluorochromes for drop-ins have minimal impact on the resolution of the backbone panel. The fluorochromes used for the backbone panel have minimal resolution impact on the detectors allocated for drop-in fluorochromes and the specified fluorochromes available for drop-in do not impact each other. The Human T Cell Backbone Panel is compatible with intracellular stain and transcription factor analysis.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Conjugate Fluorochromes (BV786, R718, PE-Cy7, BV510, BV711)
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Product Type Antibody
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.