Learn More
Raybiotech Inc C-Series Human T Cell Respose Array 1 Kit.
Supplier:  Raybiotech Inc AAHTCR18
Encompass_Preferred
Detects 10 Human proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: IL-2, IL-4, IL-5, IL-9, IL-10, IL-12 p70, IL-17A, IFN-gamma, TGF beta 1, TNF alpha
Catalog No. 50-194-8656