Learn More
Raybiotech Inc C-Series Human T Cell Response Array C2.
Supplier: Raybiotech Inc AAHTCR28
Detects 20 Human Proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: IL-2, IL-4, IL-5, IL-6, IL-7, IL-10, IL-12 p70, IL-13, IFN-gamma, TGF beta 1, TNF alpha, IL-17A, IL-9, IL-22, IL-17F, IL-18, IL-23 p19, IL-21, IL-27, IL-33 (IL-1 F11)