Learn More
American Research Products Inc Human T H glycoprotein THP EL
Catalog No. 5000328671
Encompass
Human T-H glycoprotein THP ELISA Kit from CUSABIO Detection Range 2 34 ng/mL-150 ng/mL Sensitivity 0 59 ng/mL Sample serum plasma tissue homogenates urine Method Sandwich
Catalog No. 50-003-28671 Supplier American Research Products Inc Supplier No. CSBE09451H
Human T-H glycoprotein THP ELISA Kit from CUSABIO Detection Range 2 34 ng/mL-150 ng/mL Sensitivity 0 59 ng/mL Sample serum plasma tissue homogenates urine Method Sandwich