Learn More
Proteintech Group Inc Human t-Plasminogen activator ELISA Kit
SDP
Supplier:  Proteintech Group Inc KE0018096T
Encompass_Preferred
KE00180 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The t-Plasminogen activator ELISA kit is to be used to detect and quantify protein levels of endogenous t-Plasminogen activator. The assay recognizes human t-Plasminogen activator. An antibody specific for t-Plasminogen activator has been pre-coated onto the microwells. The t-Plasminogen activator protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody specific for t-Plasminogen activator is added to detect the captured t-Plasminogen activator protein. For signal development, horseradish peroxidase (HRP)-conjugated is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
Catalog No. 50-225-8409