Learn More
ACROBiosystems ACROBiosystems Human TACI / TNFRSF13B (R119A, R122A) Protein, Fc Tag (MALS verified)
Catalog No. 502106443
Encompass_Preferred
Recombinant Protein;1MG;Human TACI (R119A, R122A), Fc Tag (TAI-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1 (R119A,R122A)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 2 μg/mL (100 μL/well) can bind Human TACI (R119A, R122A), Fc Tag (Cat. No. TAI-H5253) with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-210-6443 Supplier ACROBiosystems Supplier No. TAIH52531MG
Recombinant Protein;1MG;Human TACI (R119A, R122A), Fc Tag (TAI-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1 (R119A,R122A)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 2 μg/mL (100 μL/well) can bind Human TACI (R119A, R122A), Fc Tag (Cat. No. TAI-H5253) with a linear range of 0.2-2 ng/mL (QC tested).