Learn More
Sino Biological Human TACSTD2 Protein 100ug (Sino Biological)
Catalog No. 502371579
Encompass_Preferred
A DNA sequence encoding the Human TACSTD2 (NP_002344.2) (Met1-Thr274) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1579 Supplier Sino Biological Supplier No. 10428H41HB100
A DNA sequence encoding the Human TACSTD2 (NP_002344.2) (Met1-Thr274) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.