Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TAFA1 ELISA Kit

Catalog No. EH443RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH443RBX5 5 x 96 Tests
EH443RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH443RBX5 Supplier Invitrogen™ Supplier No. EH443RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human TAFA1 quantitates human TAFA1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TAFA1.

TAFA Chemokine Like Family Member 1 (TAFA1) is a gene located on chromosome 7q31.1, encoding a protein that is part of the FAM19A family, known for its role in modulating inflammation and neural development. TAFA1 is expressed in the brain where it influences brain-tissue homeostasis and neuronal signaling pathways. Functionally, TAFA1 acts as a chemokine-like protein, impacting the migration and adhesion of neuronal cells and contributing to neuroprotective activities. Its expression is implicated in maintaining neural network integrity and regulating processes such as neurogenesis and synaptic plasticity. Dysregulation of TAFA1 has been associated with neuroinflammatory conditions and neurodegenerative diseases, reflecting its importance in neural health and pathology. Research into TAFA1 aims to uncover its exact biological roles and potential as a therapeutic target in conditions involving disrupted cellular communication and inflammation in the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7Z5A9
Assay Range 0.12-25 ng/mL
Assay Sensitivity 0.12 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Chemokine-like protein TAFA-1; FAM19A1; family with sequence similarity 19 (chemokine (C-C motif)-like), member A1; family with sequence similarity 19 member A1, C-C motif chemokine like; protein FAM19A1; TAFA1; TAFA-1Chemokine-like protein TAFA-1; FAM19A1; family with sequence similarity 19 (chemokine (C-C motif)-like), member A1; family with sequence similarity 19 member A1, C-C motif chemokine like; protein FAM19A1; TAFA1; TAFA-1;
Gene ID (Entrez) 407738
Gene Symbol FAM19A1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target TAFA1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.