Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TAFA5/FAM19A5 ELISA Kit

Catalog No. EH445RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH445RBX5 5 x 96 Tests
EH445RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH445RBX5 Supplier Invitrogen™ Supplier No. EH445RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human TAFA5/FAM19A5 quantitates human TAFA5/FAM19A5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TAFA5/FAM19A5.

Acts as a chemokine-like protein by regulating cell proliferation and migration through activation of G protein-coupled receptors (GPCRs), such as S1PR2 and FPR2. Stimulates chemotactic migration of macrophages mediated by the MAPK3/ERK1 and AKT1 pathway. Blocks TNFSF11/RANKL-induced osteoclast formation from macrophages by inhibiting up-regulation of osteoclast fusogenic and differentiation genes. Stimulation of macrophage migration and inhibition of osteoclast formation is mediated via GPCR FPR2. Acts as an adipokine by negatively regulating vascular smooth muscle cell (VSMC) proliferation and migration in response to platelet-derived growth factor stimulation via GPCR S1PR2 and G protein GNA12/GNA13-transmitted RHOA signaling . Inhibits injury-induced cell proliferation and neointima formation in the femoral arteries.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7Z5A7
Assay Range 15-4000 pg/mL
Assay Sensitivity 15 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Chemokine-like protein TAFA-5; FAM19A5; family with sequence similarity 19 (chemokine (C-C motif)-like), member A5; family with sequence similarity 19 member A5, C-C motif chemokine like; family with sequence similarity 19, member A5; protein FAM19A5; QLLK5208; RGD1562115; TAFA protein 5; TAFA5; Tafa-5; Tara-5; UNQ5208; UNQ5208/PRO34524Chemokine-like protein TAFA-5; FAM19A5; family with sequence similarity 19 (chemokine (C-C motif)-like), member A5; family with sequence similarity 19 member A5, C-C motif chemokine like; family with sequence similarity 19, member A5; protein FAM19A5; QLLK5208; RGD1562115; TAFA protein 5; TAFA5; Tafa-5; Tara-5; UNQ5208; UNQ5208/PRO34524;
Gene ID (Entrez) 25817
Gene Symbol TAFA5
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target TAFA5
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.