Learn More
Sino Biological Human TARC/CCL17 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785459
Encompass_Preferred
The Human CCL17/TARC ELISA Pair Set is for the quantitative determination of Human CCL17/TARC.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CCL17/TARC coated on a 96-well plate. Standards and samples are added to the wells, and any Human CCL17/TARC present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CCL17/TARC monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CCL17/TARC present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85459 Supplier Sino Biological Supplier No. SEKA102335
The Human CCL17/TARC ELISA Pair Set is for the quantitative determination of Human CCL17/TARC.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CCL17/TARC coated on a 96-well plate. Standards and samples are added to the wells, and any Human CCL17/TARC present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CCL17/TARC monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CCL17/TARC present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.