Learn More
American Research Products Inc Human Tartrate Resistant Acid
Catalog No. 5000328326
Encompass
Human Tartrate Resistant Acid Phosphatase (TRAP) ELISA Kit from CUSABIO Detection Range 0 47 ng/mL-30 ng/mL Sensitivity 0 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28326 Supplier American Research Products Inc Supplier No. CSBE13415H
Human Tartrate Resistant Acid Phosphatase (TRAP) ELISA Kit from CUSABIO Detection Range 0 47 ng/mL-30 ng/mL Sensitivity 0 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich