Learn More
Biomatik Corporation Human TATA Box Binding Protein Associated Factor 13 (TAF13) ELISA Kit
Supplier:  Biomatik Corporation EKU07574
Encompass_Preferred
Human TATA Box Binding Protein Associated Factor 13 (TAF13) ELISA Kit for detecting TAF13 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.128ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15543. Gene ID: 6884. Target Synonyms: TAF2K; TAFII18; Transcription Initiation Factor TFIID Subunit 13; Transcription initiation factor TFIID 18 kDa subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5721