Learn More
ACROBiosystems Human Tau 500ug
Supplier:  ACROBiosystems TAUH5147500UG
Encompass_Preferred
Human Tau-441 GSK-3beta-phosphorylated protein His Tag (TAU-H5147) is expressed from E. coli cells. It contains AA Met 1 - Leu 441 (Accession # P10636-8).
Catalog No. 50-253-4073