Learn More
Bon Opus Biosciences Recombinant Human Tau protein - Source: E.coli
SDP
Supplier:  Bon Opus Biosciences BP22750UG
Encompass_Preferred
Recombinant Human Tau is produced by our E.coli expression system and the target gene encoding Met1-Leu441 is expressed. Accession#P10636-8. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM EDTA, pH 7.5
Catalog No. NC2733667