Learn More
Raybiotech Inc Human MAPT (Tau) ELISA Kit
Supplier:  Raybiotech Inc ELHTAU1
Encompass_Preferred
For cell culture supernatants, EDTA and/or Citrate treated plasma, and serum samples. Heparin is not recommended. Sensitivity: 100 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Tau. Other species not determined.
Catalog No. 50-194-9762