Learn More
Abcam Human Tau ELISA Kit, 1 x 96 Tests, Colorimetric, Sandwich ELISA (quantitative), Detection Range 40.63-2600 pg/mL, Sensitivity 16 pg/mL, Assay Time 90 min
Supplier: Abcam AB2736171X96TESTS
The Abcam Human Tau ELISA Kit is a rapid, single-wash SimpleStep ELISA designed for quantitative measurement of human Tau protein, suitable for research into neurological conditions such as Alzheimer's disease. The kit is noted for its high sensitivity and specificity, allowing researchers to detect Tau in various biological samples.
- Detection method: Colorimetric
- Assay type: Sandwich ELISA (quantitative)
- Assay platform: Pre-coated 96-well microplate
- Assay time: 90 min
- Sensitivity: 16 pg/mL
- Detection range: 40.63 - 2600 pg/mL
- Reacts with: Human
- Sample types: Cell culture extracts, Tissue extracts, Cell culture media, Heparin plasma, Serum, EDTA plasma, Cerebral spinal fluid (CSF)
- Specificity: Detects native and recombinant human Tau protein
- Cross-reactivity: None with UCHL-1 or alpha synuclein
- Stability: 1 year at 4°C if unopened
- Product size: 1 x 96 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.