Learn More
Bon Opus Biosciences Recombinant Human Tau protein (N-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP22850UG
Encompass_Preferred
Recombinant Human Tau protein is produced by our Mammalian expression system and the target gene encoding Ala2-Leu441 is expressed with a 6His tag at the N-terminus. Accession# P10636-8. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP228
Catalog No. 50-217-2714