Learn More
Abcam Human Tau (phospho T217) Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502699161
Encompass_Preferred
Human Tau (phospho T217) Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Tau (phospho T217).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-9161 Supplier Abcam Supplier No. AB31455710X96TESTS
Human Tau (phospho T217) Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Tau (phospho T217).
The product is subject to the following: Abcam Restricted Use Statement