Learn More
Biomatik Corporation Human Tau Protein (MAPT) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12190
Encompass_Preferred
Human Tau Protein (MAPT) Mini sample ELISA Kit for detecting MAPT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10636. Gene ID: 4137. Target Synonyms: DDPAC; FTDP-17; MAPTL; MSTD; MTBT1; MTBT2; PPND; Neurofibrillary tangle protein; Microtubule Associated Protein Tau; Paired helical filament-tau; G Protein Beta1/Gamma2 Subunit-Interacting Factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8011