Learn More
Meso Scale Discovery Human Tau (pT181) Kit, 96-well plate, for neurobiology research, single-analyte, sensitivity 77 fg/mL, dynamic range 77 - 990,000 fg/mL, sample types: serum, plasma, CSF, cell culture supernatant, urine
SDP
Catalog No. NC2744835
Encompass
Human Tau (pT181) Kit, 96-well plate, for neurobiology research, single-analyte, sensitivity 77 fg/mL, dynamic range 77 - 990,000 fg/mL, sample types: serum, plasma, CSF, cell culture supernatant, urine
Catalog No. NC2744835 Supplier Meso Scale Discovery Supplier No. K151AGMS1
Human Tau (pT181) Kit, 96-well plate, for neurobiology research, single-analyte, sensitivity 77 fg/mL, dynamic range 77 - 990,000 fg/mL, sample types: serum, plasma, CSF, cell culture supernatant, urine