Learn More
Biomatik Corporation Human Taurine Transporter (TAUT) ELISA Kit
Supplier:  Biomatik Corporation EKU09191
Encompass_Preferred
Human Taurine Transporter (TAUT) ELISA Kit for detecting TAUT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31641. Gene ID: 6533. Target Synonyms: SLC6A6; Sodium And Chloride Dependent Taurine Transporter; Solute Carrier Family 6 Member 6,Neurotransmitter Transporter,Taurine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8553