Learn More
Biomatik Corporation Human TBC1 Domain Family, Member 13 (TBC1D13) ELISA Kit
Supplier:  Biomatik Corporation EKU09380
Encompass_Preferred
Human TBC1 Domain Family, Member 13 (TBC1D13) ELISA Kit for detecting TBC1D13 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NVG8. Gene ID: 54662. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8530