Learn More
Biomatik Corporation Human T-Box Protein 21 (TBX21) ELISA Kit
Supplier:  Biomatik Corporation EKU07579
Encompass_Preferred
Human T-Box Protein 21 (TBX21) ELISA Kit for detecting TBX21 in cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.052ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UL17. Gene ID: 30009. Target Synonyms: TBLYM; TBET; T-box transcription factor TBX21; T-cell-specific T-box transcription factor T-bet. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5598