Learn More
Sino Biological Anti-TBX6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105152T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TBX6, and purified by antigen affinity chromatography.
Catalog No. 50-241-1870