Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TCCR (aa 40-122) Control Fragment Recombinant Protein

Catalog No. RP109344
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109344 Supplier Invitrogen™ Supplier No. RP109344
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

In mice, CD4+ helper T-cells differentiate into type 1 (Th1) cells and type 2 (Th2) cells, which are critical for cell-mediated immunity and most antibody responses, respectively. The differentiation of Th1 cells is predominantly under the influence of IL12, while IL4 influences the differentiation of Th2 cells. Mice deficient in these cytokines, their receptors, or associated transcription factors have impaired, but not absent, Th1 or Th2 immune responses. The gene discussed here encodes a protein that is similar to the mouse T-cell cytokine receptor Tccr at the amino acid level and is predicted to be a glycosylated transmembrane protein. Diseases associated with IL27RA include Hyper Ige Recurrent Infection Syndrome 4 and Crisponi/Cold-Induced Sweating Syndrome 2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6UWB1
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 9466
Name Human TCCR (aa 40-122) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias class I cytokine receptor; CRL1; CRL1IL-27R; cytokine receptor family, class 1 (WSXWS), member 1; cytokine receptor WSX-1; Cytokine receptor-like 1; il-27 receptor; IL-27 receptor subunit alpha; IL27R; IL-27R; IL-27R subunit alpha; IL27RA; IL-27RA; IL-27R-alpha; interleukin 27 receptor subunit alpha; interleukin 27 receptor, alpha; interleukin-27 receptor subunit alpha; T cell cytokine receptor; T cell cytokine receptor; cytokine receptor family, class 1 (WSXWS), member 1; TCCR; T-cell cytokine receptor type 1; Type I T-cell cytokine receptor; UNQ296/PRO336; WSX1; WSX-1; ZcytoR1
Common Name TCCR
Gene Symbol Il27ra
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QCYGVGPLGDLNCSWEPLGDLGAPSELHLQSQKYRSNKTQTVAVAAGRSWVAIPREQLTMSDKLLVWGTKAGQPLWPPVFVNL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.