Learn More
Sino Biological Recombinant Human Cripto Protein (His Tag)
Supplier:  Sino Biological 10908H08H50
Encompass_Preferred
A DNA sequence encoding the human TDGF1 (AAH22393.1) (Met 1-Thr 172) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-237-4473