Learn More
Proteintech Group Inc Human TDP-43 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0000596T
Encompass_Preferred
TARDBP was firstly found as a transcriptional repressor that binds to chromosomally integrated TAR DNA and represses HIV-1 transcription. It was also reported to regulate alternate splicing of the CFTR gene. Neumann et al. (2006) found that a hyperphosphorylated, ubiquitinated, and cleaved form of TARDBP, known as pathologic TDP43, is the major disease protein in ubiquitin-positive, tau-, and alpha-synuclein-negative frontotemporal dementia (FTLD-U) and in Amyotrophic lateral sclerosis (ALS). This kit is used to quantify TDP-43 level in vivo.
Catalog No. 50-197-3988