Learn More
ACROBiosystems Human TDP-43, His Tag (TD3-H5145) is expressed from E.coli cells. It contains AA Met 1 - Met 414 (Accession # Q13148-1).
Supplier:  ACROBiosystems TD3H5145500UG
Encompass_Preferred
TDP-43, the transactive response (TAR)-DNA binding Protein with a molecular weight of 43 KDa is encoded by TARDBP gene that located at the chromosome 1. It is a RNA/DNA binding protein that structurally resembles a typical hnRNP protein family member. involved in transcriptional regulation and RNA processing. It is linked to sporadic and familial amyotrophic lateral sclerosis and frontotemporal lobar degeneration. TDP-43 is predominantly nuclear, but it translocates to the cytoplasm under pathological conditions. Cytoplasmic accumulation, phosphorylation, ubiquitination and truncation of TDP-43 are the main hallmarks of TDP-43 proteinopathies.
Catalog No. 50-225-3881