Learn More
Biomatik Corporation Human TEA Domain Family, Member 1 (TEAD1) ELISA Kit
Supplier:  Biomatik Corporation EKU12503
Encompass_Preferred
Human TEA Domain Family, Member 1 (TEAD1) ELISA Kit for detecting TEAD1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28347. Gene ID: 7003. Target Synonyms: REF1; AA; TCF13; TEF-1; NTEF-1; Transcription factor 13; Atrophia Areata,Peripapillary Chorioretinal Degeneration; SV40 Transcriptional Enhancer Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5932