Learn More
Sino Biological Human TEK MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10700MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TIE2 (rh TIE2; Catalog#10700-H08H; AAA61139.1; Met 1-Lys 745). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2608