Learn More
Sino Biological Recombinant Anti-TIE-2 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10700R116A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Tie2 / CD202b / TEK (rh Tie2 / CD202b / TEK; Catalog#10700-H08H; AAA61139.1; Met 1-Lys 745) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2127