Learn More
Sino Biological Anti-TIE-2 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10700MM07A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Tie2 / CD202b / TEK (rh Tie2 / CD202b / TEK; Catalog#10700-H08H; AAA61139.1; Met1-Lys745) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-1330