Learn More
Sino Biological Human TEK MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10700R11620
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Tie2 / CD202b / TEK (rh Tie2 / CD202b / TEK; Catalog#10700-H08H; AAA61139.1; Met1-Lys745).
Catalog No. 50-237-0616