Learn More
Sino Biological Anti-TIE-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10700T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TIE-2 (rh TIE-2; Catalog#10700-H08H; AAA61139.1; Met1-Lys745). TIE-2 specific IgG was purified by Human TIE-2 affinity chromatography.
Catalog No. 50-255-5196